Northern Ireland
Order Entry
Anti-C3 Goat Polyclonal Antibody
Anti-C3 Goat Polyclonal Antibody
  BIRBORB25831-10
 :  Biorbyt
Anti-C3 Goat Polyclonal Antibody
  BIRBORB25831-10
 :  Biorbyt
 
 
   
  1 * 10 mL

 

  • Antibody type:
    Primary
  • Antigen name:
    complement component 3
  • Antigen symbol:
    C3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Storage buffer:
    0,05M Tris, 0,5M Sodium Chloride, pH 7,5.
  • Storage temperature:
    Cool
  • Concentration:
    Total Protein: 53 OD280 Units/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified Human C3
  • Purification:
    purified by delipidation followed by solid phase adsorption
  • Size:
    10 ml
  • Pk:
    10 mL

 

 

Type: Primary
Antigen: C3 (complement component 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: