Anti-MPO Mouse Monoclonal Antibody
Catalog # BIRBORB25561-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Myeloperoxidase
- Antigen symbol:MPO
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:4,2 mg/ml (Sigma protein assay kit)
- Shipping temperature:Ice
- Immunogen:Human myeloperoxidase purified leukocyte mass
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: