Anti-PRTN3 Mouse Monoclonal Antibody
BIRBORB25560-1
: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:proteinase 3
- Antigen symbol:PRTN3
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:6,4 mg/ml (Sigma Protein Assay Kit)
- Shipping temperature:Ice
- Immunogen:Human proteinase 3 (PR3) purified leukocyte mass.
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
Type: Primary
Antigen: PRTN3 (proteinase 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human