Northern Ireland
Order Entry
Anti-EGFR, Activated Mouse monoclonal antibody
  BIRBORB25523-100
 :  Biorbyt
undefined
Anti-EGFR, Activated Mouse monoclonal antibody
  BIRBORB25523-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    EGFR activated
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized
  • Modification:
    phospho-Tyr 1173
  • Storage temperature:
    Cool
  • Concentration:
    100 ug/ml (prior to lyophilization)
  • Shipping temperature:
    Ice
  • Immunogen:
    Phosphopeptide conjugated to KLH
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

DB: 0.5 ?g/ml; Immunofluorescence: 1-10 ?g/ml; ELISA: 0.05 ?g/ml

Type: Primary
Antigen: EGFR, Activated
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human