Anti-Relaxin-2 Rabbit Polyclonal Antibody
BIRBORB25195-10
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Relaxin-2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Form:lyophilized
- Storage buffer:50mM Tris, pH 7,5
- Storage temperature:Cool
- Concentration:1 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Purified human Relaxin-2
- Purification:purified by Protein G chromatography
- Size:10 µg
- Pk:10 µG
ELISA: 1 ?g/ml; RIA: 1 ?g/ml; Immunofluorescence: 5 ?g/ml
Type: Primary
Antigen: Relaxin-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: