Northern Ireland
Order Entry
Anti-ALB Goat Polyclonal Antibody
  BIRBORB25141-10
 :  Biorbyt
Anti-ALB Goat Polyclonal Antibody
Catalog # BIRBORB25141-10
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 10 mL

 

  • Antibody type:
    Primary
  • Antigen name:
    albumin
  • Antigen symbol:
    ALB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Reactivity:
    Human
  • Storage buffer:
    0,05M Tris-HC1, pH 7,5 containing 0,5M Sodium chloride
  • Storage temperature:
    Cool
  • Concentration:
    Total protein: Lot Specific g/dl,Titer: Lot Specific mg/ml (Becker titer)
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified Human Albumin
  • Purification:
    purified by defibrination, delipidation and absorbed by solid phase chromatography
  • Size:
    10 ml
  • Pk:
    10 mL

 

About this item

Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human