Anti-Hemagglutinin Peptide Rabbit Polyclonal Antibody
BIRBORB25116-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Hemagglutinin Peptide
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:1 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:YPYDVPDYA (Influenza hemagglutinin-HA-epitope) conjugated to KLH
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:200-1:500; Western Blot: 1:5000-1:10000; Immunoprecipitation: 1:250-1:2000
Type: Primary
Antigen: Hemagglutinin Peptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: