Anti-proBNP Mouse Monoclonal Antibody
Catalog # BIRBORB24977-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:proBNP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:N-terminal
- Amino acid number:13 to 27
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:5,6 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Synthetic NT-proBNP peptide corresponding to amino acids 13-27
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
Type: Primary
Antigen: proBNP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human