Anti-LTF Mouse Monoclonal Antibody [clone: 1A1]
BIRBORB24830-1
: Biorbyt
- Antibody type:Primary
- Antigen name:lactotransferrin
- Antigen symbol:LTF
- Clonality:Monoclonal
- Clone:1A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:4,3 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Purified lactoferrin from human milk
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: monoclonal
Clone: 1A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human