Anti-M-Cadherin Mouse Monoclonal Antibody
BIRBORB24812-100
: Biorbyt
- Antibody type:Primary
- Antigen name:M-Cadherin
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Storage buffer:2X PBS, PEG and Sucrose
- Storage temperature:Cool
- Concentration:100 ug/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Recombinant extracellular domain of human M-Cadherin
- Purification:purified by Thiophilic adsorption and size exclusion chromatography
- Size:100 µg
- Pk:100 µG
About this item
DB: 1 ?g/ml; ELISA: 0.05 ?g/ml; Immunofluorescence: 5 ?g/ml
Type: Primary
Antigen: M-Cadherin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human