Northern Ireland
Order Entry
Anti-Methadone Mouse Monoclonal Antibody
Anti-Methadone Mouse Monoclonal Antibody
  BIRBORB24615-1
 :  Biorbyt
Anti-Methadone Mouse Monoclonal Antibody
  BIRBORB24615-1
 :  Biorbyt
 
 
   
  1 * 1 mg

 

  • Antibody type:
    Primary
  • Antigen name:
    Methadone
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG
  • Storage buffer:
    0,05 M Potassium Phosphate, 0,85% Sodium Chloride, pH 7.15.
  • Storage temperature:
    Cool
  • Concentration:
    2,89 mg/ml (OD280)
  • Shipping temperature:
    Ice
  • Immunogen:
    Methadone-BSA
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Methadone
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: