Anti-Oxycodone Mouse Monoclonal Antibody
BIRBORB24522-1
: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Oxycodone
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Storage buffer:PBS, pH 7.0
- Storage temperature:Cool
- Concentration:(OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Oxycodone-BSA conjugate
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Oxycodone
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: