Northern Ireland
Order Entry
Anti-PEG Mouse Monoclonal Antibody
Anti-PEG Mouse Monoclonal Antibody
  BIRBORB24460-100
 :  Biorbyt
Anti-PEG Mouse Monoclonal Antibody
  BIRBORB24460-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PEG
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by HPLC
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: PEG
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: