Anti-hCG beta Mouse monoclonal antibody
Catalog # BIRBORB24433-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:HCG beta
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:6 mg/ml (Sigma Protein Assay Kit)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: hCG beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human