Anti-Thyroxine (T4) Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB24390-1
Supplier: Biorbyt
- Antibody type:Primary
- Antigen name:Thyroxine (T4)
- Clonality:Monoclonal
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1
- Storage buffer:20mM Tris-HCL, 150mM Sodium chloride, pH 7,5.
- Storage temperature:Cool
- Concentration:1,3 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Conjugate of purified thyroxine with BSA.
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
About this item
Type: Primary
Antigen: Thyroxine (T4)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: