Anti-hCG alpha Mouse monoclonal antibody
Catalog # BIRBORB24365-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:HCG alpha
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Storage buffer:0,15M Sodium chloride, 10mM Tris, pH 7,5
- Storage temperature:Cool
- Concentration:2,4 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Purified hCG alpha chain.
- Purification:purified by ion exchange chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: hCG alpha
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human