Northern Ireland
Order Entry
Anti-Estriol Mouse Monoclonal Antibody
Anti-Estriol Mouse Monoclonal Antibody
  BIRBORB24354-1
 :  Biorbyt
Anti-Estriol Mouse Monoclonal Antibody
  BIRBORB24354-1
 :  Biorbyt
 
 
   
  1 * 1 mg

 

  • Antibody type:
    Primary
  • Antigen name:
    Estriol
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Concentration:
    4,61 mg/ml (OD280)
  • Shipping temperature:
    Ice
  • Immunogen:
    3-CME-Estriol-BgG
  • Purification:
    purified
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Estriol
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: