Northern Ireland
Order Entry
Anti-HCV NS-3 Mouse monoclonal antibody
Anti-HCV NS-3 Mouse monoclonal antibody
  BIRBORB24157-100
 :  Biorbyt
Anti-HCV NS-3 Mouse monoclonal antibody
  BIRBORB24157-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HCV NS-3
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Amino acid number:
    1350 to 1460
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant antigen (HCV genotype 1b)
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

ELISA: 1:20

Type: Primary
Antigen: HCV NS-3
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: