Anti-Astrovirus Mouse monoclonal antibody
BIRBORB23981-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Astrovirus
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Western blot:Yes
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:2,3 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Inactivated Astrovirus, Serotype 1.
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Astrovirus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: