Anti-Diphtheria Toxin Mouse monoclonal antibody
BIRBORB23968-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Diphtheria Toxin
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:8,6 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Diphtheria toxoid.
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Diphtheria Toxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: