Northern Ireland
Order Entry
Anti-Epstein Barr Virus Mouse Monoclonal Antibody (FITC (Fluorescein))
Anti-Epstein Barr Virus Mouse Monoclonal Antibody (FITC (Fluorescein))
  BIRBORB23800-100
 :  Biorbyt
Anti-Epstein Barr Virus Mouse Monoclonal Antibody (FITC (Fluorescein))
  BIRBORB23800-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Epstein Barr Virus
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Storage buffer:
    0,01 M PBS, pH 7,2 containing 10 mg/mL BSA
  • Storage temperature:
    Cool
  • Concentration:
    100 ug/ml (OD280, E(0,1%)=1,3)
  • Shipping temperature:
    Ice
  • Immunogen:
    Infected B cell Lysate (Native Protein).
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: Epstein Barr Virus
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: