Northern Ireland
Order Entry
Anti-MRSA Mouse Monoclonal Antibody
Anti-MRSA Mouse Monoclonal Antibody
Catalog # BIRBORB23480-0.5
Supplier:  Biorbyt
Anti-MRSA Mouse Monoclonal Antibody
Catalog # BIRBORB23480-0.5
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 0,5 mg

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MRSA
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Form:
    lyophilized
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml (OD280, E(0,1%)=1,41) (prior to lyophilization)
  • Shipping temperature:
    Ice
  • Immunogen:
    Gel purified MRSA protein (penicillin-binding protein 2 from strain EMRSA-16)
  • Purification:
    purified by Protein G chromatography
  • Size:
    500 μg
  • Pk:
    0,5 mg

Specifications

About this item

ELISA: 1:5000; DB: 1:2000

Type: Primary
Antigen: MRSA
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: