Anti-M. tuberculosis ESAT6 Mouse Monoclonal Antibody
Catalog # BIRBORB23449-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:M. tuberculosis ESAT6
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:2,0 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Recombinant protein ESAT6 M. tuberculosis H37RV
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: M. tuberculosis ESAT6
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: