Northern Ireland
Order Entry
Anti-Influenza A H5 Mouse Monoclonal Antibody
  BIRBORB23384-1
 :  Biorbyt
Anti-Influenza A H5 Mouse Monoclonal Antibody
  BIRBORB23384-1
 :  Biorbyt
 
 
   
  1 * 1 mg

 

  • Antibody type:
    Primary
  • Antigen name:
    Influenza A H5
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    3,5 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified avian influenza virus type A (H5N1)
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Influenza A H5
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: