Anti-Influenza A H5N1 Goat Polyclonal Antibody
BIRBORB23306-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Influenza A H5N1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide corresponding to 14 amino acids near the middle of the Hemagglutinin protein
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Influenza A H5N1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: