Northern Ireland
Order Entry
Anti-Salmonella sp. Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-Salmonella sp. Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BIRBORB23227-1
 :  Biorbyt
Anti-Salmonella sp. Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB23227-1
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 1 mL

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Salmonella sp.
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Storage buffer:
    PBS containing 10mg/ml BSA
  • Storage temperature:
    Cool
  • Concentration:
    1-2 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Mixture of S. enteriditis, S. typhimurium and S. heidelburg
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Primary
Antigen: Salmonella sp.
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: