Anti-Vaccinia Virus Rabbit Polyclonal Antibody (Biotin)
BIRBORB23077-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Vaccinia Virus
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Storage buffer:0,01M PBS, pH 7,2Product does not contain stabilizing proteins
- Storage temperature:Cool
- Concentration:4-5 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Lister Strain (mixture of virions and infected cell polypeptides).
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Vaccinia Virus
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: