Northern Ireland
Order Entry
Anti-NCR3 Mouse Monoclonal Antibody
Anti-NCR3 Mouse Monoclonal Antibody
Supplier:  Biorbyt
Certificates
Anti-NCR3 Mouse Monoclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22889-20
  • BIRBORB22889-5
  • BIRBORB22889-100
  • Antigen symbol:
  • NCR3
  • NCR3
  • NCR3
  • Antigen name:
  • Natural cytotoxicity triggering receptor 3
  • Natural cytotoxicity triggering receptor 3
  • Natural cytotoxicity triggering receptor 3
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Anti-human NKp30 mAb, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c immunized with recombinant human NKp30 amino acids 19-138 purified from E.coli
  • Anti-human NKp30 mAb, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c immunized with recombinant human NKp30 amino acids 19-138 purified from E.coli
  • Anti-human NKp30 mAb, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c immunized with recombinant human NKp30 amino acids 19-138 purified from E.coli
  • Amino acid number:
  • 19 to 138
  • 19 to 138
  • 19 to 138
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BIRBORB22889-20
    ORB22889-20
    20 µg
    20 µG
    Log in for availability
    Log in for availability
    1 * 20 µG
    1 * 20 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    NCR3
    Antigen name
    Natural cytotoxicity triggering receptor 3
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Anti-human NKp30 mAb, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c immunized with recombinant human NKp30 amino acids 19-138 purified from E.coli
    Amino acid number
    19 to 138
    Purification
    purified by Protein G chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB22889-5
    ORB22889-5
    5 µg
    5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    NCR3
    Antigen name
    Natural cytotoxicity triggering receptor 3
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Anti-human NKp30 mAb, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c immunized with recombinant human NKp30 amino acids 19-138 purified from E.coli
    Amino acid number
    19 to 138
    Purification
    purified by Protein G chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB22889-100
    ORB22889-100
    100 µg
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    NCR3
    Antigen name
    Natural cytotoxicity triggering receptor 3
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Anti-human NKp30 mAb, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c immunized with recombinant human NKp30 amino acids 19-138 purified from E.coli
    Amino acid number
    19 to 138
    Purification
    purified by Protein G chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice