Northern Ireland
Order Entry
Anti-GMNN Mouse monoclonal antibody
Anti-GMNN Mouse monoclonal antibody
 :  Biorbyt
 
Anti-GMNN Mouse monoclonal antibody
 :  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22828-5
  • BIRBORB22828-20
  • BIRBORB22828-100
  • Antigen symbol:
  • GMNN
  • GMNN
  • GMNN
  • Antigen name:
  • geminin, DNA replication inhibitor
  • geminin, DNA replication inhibitor
  • geminin, DNA replication inhibitor
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Anti-human Geminin mAb is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human Geminin amino acids 1-209 purified from E.coli
  • Anti-human Geminin mAb is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human Geminin amino acids 1-209 purified from E.coli
  • Anti-human Geminin mAb is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human Geminin amino acids 1-209 purified from E.coli
  • Amino acid number:
  • 1 to 209
  • 1 to 209
  • 1 to 209
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BIRBORB22828-5
    ORB22828-5
    5 µg
    5 µG
       
    Log in for availability
    1 * 5 µG
    1 * 5 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    GMNN
    Antigen name
    geminin, DNA replication inhibitor
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Anti-human Geminin mAb is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human Geminin amino acids 1-209 purified from E.coli
    Amino acid number
    1 to 209
    Purification
    purified by Protein G chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB22828-20
    ORB22828-20
    20 µg
    20 µG
       
    Log in for availability
    1 * 20 µG
    1 * 20 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    GMNN
    Antigen name
    geminin, DNA replication inhibitor
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Anti-human Geminin mAb is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human Geminin amino acids 1-209 purified from E.coli
    Amino acid number
    1 to 209
    Purification
    purified by Protein G chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB22828-100
    ORB22828-100
    100 µg
    100 µG
       
    Log in for availability
    1 * 100 µG
    1 * 100 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    GMNN
    Antigen name
    geminin, DNA replication inhibitor
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Anti-human Geminin mAb is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human Geminin amino acids 1-209 purified from E.coli
    Amino acid number
    1 to 209
    Purification
    purified by Protein G chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice