Northern Ireland
Order Entry
Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein))
 :  Biorbyt
 
Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein))
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB22653-500
  • BIRBORB22653-1
  • Antigen symbol:
  • CD14
  • CD14
  • Antigen name:
  • CD14 molecule
  • CD14 molecule
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Purified human Cells.
  • Purified human Cells.
  • Purification:
  • purified by ion exchange chromatography
  • purified by ion exchange chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • BIRBORB22653-500
    ORB22653-500
    500 µg
    500 µG
       
       
    1 * 500 µG
    1 * 500 µG

     
     
    Specifications
    Antigen symbol
    CD14
    Antigen name
    CD14 molecule
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    FITC (Fluorescein Isothiocyanate)
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Purified human Cells.
    Purification
    purified by ion exchange chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB22653-1
    ORB22653-1
    1 mg
    1 mg
       
       
    1 * 1 mg
    1 * 1 mg

     
     
    Specifications
    Antigen symbol
    CD14
    Antigen name
    CD14 molecule
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    FITC (Fluorescein Isothiocyanate)
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Immunogen
    Purified human Cells.
    Purification
    purified by ion exchange chromatography
    Storage temperature
    Cool
    Shipping temperature
    Ice