Northern Ireland
Order Entry
Anti-IgE Sheep Polyclonal Antibody
Catalog # BIRBORB22438-1
Supplier:  Biorbyt
Anti-IgE Sheep Polyclonal Antibody
Catalog # BIRBORB22438-1
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 1 mL

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgE
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Format:
    antiserum
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Highly purified monoclonal IgE isolated from human serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: