Northern Ireland
Order Entry
Anti-IgM Goat Polyclonal Antibody (Biotin)
  BIRBORB21821-1
 :  Biorbyt
Anti-IgM Goat Polyclonal Antibody (Biotin)
  BIRBORB21821-1
 :  Biorbyt
 
 
   
  1 * 1 mL

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Goat
  • Format:
    immunoconjugate
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Concentration:
    7,5 mg/ml, Biotin/IgG protein molar ratio (B/P) is approximately 5,3, No foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified normal IgM isolated from pooled human serum. Immunization with intact (19S) and split IgM (7S). Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: