Northern Ireland
Order Entry
Anti-IgG Rabbit Polyclonal Antibody
Anti-IgG Rabbit Polyclonal Antibody
  BIRBORB21510-10
 :  Biorbyt
Anti-IgG Rabbit Polyclonal Antibody
  BIRBORB21510-10
 :  Biorbyt
 : ORB21510-10
 : BIRBORB21510-10
 
 
   
  1 * 10 mg

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Format:
    purified hyperimmune IgG
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified normal IgG isolated from pooled guinea pig serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    10 mg
  • Pk:
    10 mg

 

 

Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: