Anti-GAPDH Sheep Polyclonal Antibody
BIRBORB21139-10
: Biorbyt
- Antibody type:Primary
- Antigen name:Glyceraldehyde-3-phosphate dehydrogenase
- Antigen symbol:GAPDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Reactivity:Rabbit
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Glyceraldehyde-3-phosphate dehydrogenase isolated and purified from rabbit muscle. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Rabbit