Northern Ireland
Order Entry
Anti-Phospholipase D Rabbit Polyclonal Antibody (Biotin)
Anti-Phospholipase D Rabbit Polyclonal Antibody (Biotin)
  BIRBORB20940-1
 :  Biorbyt
Anti-Phospholipase D Rabbit Polyclonal Antibody (Biotin)
  BIRBORB20940-1
 :  Biorbyt
 
 
   
  1 * 1 mL

 

  • Antibody type:
    Primary
  • Antigen name:
    Phospholipase D
  • Antigen symbol:
    PLD
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • Reactivity:
    Plant
  • Format:
    immunoconjugate
  • Storage buffer:
    phosphate buffered saline
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, Biotin/IgG protein molar ratio (B/P) approximately 5,5, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    Phospholipase D isolated and purified from cabbage. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Primary
Antigen: Phospholipase D
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Plant