Anti-Ascorbate Oxidase Rabbit Polyclonal Antibody
BIRBORB20746-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Ascorbate Oxidase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Plant
- Environmentally Preferable:
- Form:lyophilized
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:1 mg/0,5ml, no foreign proteins added
- Shipping temperature:Ice
- Immunogen:Ascorbate oxidase isolated and purified from cucurbita species. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Ascorbate Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Plant