Northern Ireland
Order Entry
Anti-ALDH9A1 Goat Polyclonal Antibody
  BIRBORB20592-100
 :  Biorbyt
Anti-ALDH9A1 Goat Polyclonal Antibody
  BIRBORB20592-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    aldehyde dehydrogenase 9 family, member A1
  • Antigen symbol:
    ALDH9A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Western blot:
    Yes
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA._x000D_
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide (Internal Region)
  • Purification:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: ALDH9A1 (aldehyde dehydrogenase 9 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: