Anti-MLX Goat Polyclonal Antibody
BIRBORB20400-100
: Biorbyt
- Antibody type:Primary
- Antigen name:MLX, MAX dimerization Protein
- Antigen symbol:MLX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:128000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: MLX (MLX, MAX dimerization protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: