Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin))
: Biorbyt
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- BIRBORB12152-100
- BIRBORB12152-50
- Antigen symbol:
- HLA-DR
- HLA-DR
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage buffer:
- Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
- Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
Specifications
Antigen symbolHLA-DREnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationPE (Phycoerythrin)ReactivityHumanHostMouseIsotypeIgG1ImmunoFluorescenceYesFlow cytometryYesPurificationpurified by affinity chromatographyStorage bufferPhosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSAStorage temperatureCoolShipping temperatureIceSpecifications
Antigen symbolHLA-DREnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationPE (Phycoerythrin)ReactivityHumanHostMouseIsotypeIgG1ImmunoFluorescenceYesFlow cytometryYesPurificationpurified by affinity chromatographyStorage bufferPhosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSAStorage temperatureCoolShipping temperatureIce