Northern Ireland
Order Entry
Anti-APP Mouse polyclonal antibody
Anti-APP Mouse polyclonal antibody
 :  Biorbyt
 
undefined
Anti-APP Mouse polyclonal antibody
 :  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB10118-200
  • BIRBORB10118-100
  • Antigen symbol:
  • APP
  • APP
  • Antigen name:
  • Amyloid Beta (A4) Precursor Protein
  • Amyloid Beta (A4) Precursor Protein
  • Antigen synonyms:
  • ABETA|beta Amyloid
  • ABETA|beta Amyloid
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide
  • Synthetic peptide
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

 

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BIRBORB10118-200
    ORB10118-200
    200 µg
    200 µG
       
    Log in for availability
    1 * 200 µG
    1 * 200 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    APP
    Antigen name
    Amyloid Beta (A4) Precursor Protein
    Environmentally Preferable
    Antigen synonyms
    ABETA|beta Amyloid
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Mouse
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Immunogen
    Synthetic peptide
    Form
    liquid
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB10118-100
    ORB10118-100
    100 µg
    100 µG
       
    Log in for availability
    1 * 100 µG
    1 * 100 µG

    Specifications:
    More
    Specifications
    Antigen symbol
    APP
    Antigen name
    Amyloid Beta (A4) Precursor Protein
    Environmentally Preferable
    Antigen synonyms
    ABETA|beta Amyloid
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Mouse
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Immunogen
    Synthetic peptide
    Form
    liquid
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice