Northern Ireland
Order Entry
Anti-BMX Rabbit Polyclonal Antibody
Anti-BMX Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-BMX Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB5199-100
  • BIRBORB5199-20
  • BIRBORB5199-5
  • Antigen symbol:
  • BMX
  • BMX
  • BMX
  • Antigen name:
  • BMX non-receptor tyrosine kinase
  • BMX non-receptor tyrosine kinase
  • BMX non-receptor tyrosine kinase
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Modification:
  • phospho-Tyr556
  • phospho-Tyr556
  • phospho-Tyr556
  • Immunogen:
  • KLH conjugated synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr556.
  • KLH conjugated synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr556.
  • KLH conjugated synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr556.
  • Form:
  • liquid
  • liquid
  • liquid
  • Molecular weight:
  • 78 kDa
  • 78 kDa
  • 78 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • BIRBORB5199-100
    ORB5199-100
    100 µg
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG

     
     
    Specifications
    Antigen symbol
    BMX
    Antigen name
    BMX non-receptor tyrosine kinase
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Modification
    phospho-Tyr556
    Immunogen
    KLH conjugated synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr556.
    Form
    liquid
    Molecular weight
    78 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB5199-20
    ORB5199-20
    20 µg
    20 µG
       
       
    1 * 20 µG
    1 * 20 µG

     
     
    Specifications
    Antigen symbol
    BMX
    Antigen name
    BMX non-receptor tyrosine kinase
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Modification
    phospho-Tyr556
    Immunogen
    KLH conjugated synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr556.
    Form
    liquid
    Molecular weight
    78 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB5199-5
    ORB5199-5
    5 µg
    5 µG
       
       
    1 * 5 µG
    1 * 5 µG

     
     
    Specifications
    Antigen symbol
    BMX
    Antigen name
    BMX non-receptor tyrosine kinase
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Modification
    phospho-Tyr556
    Immunogen
    KLH conjugated synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr556.
    Form
    liquid
    Molecular weight
    78 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice