Northern Ireland
Order Entry
Anti-ERP29 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-ERP29 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB5193-100
  • BIRBORB5193-200
  • Antigen symbol:
  • ERP29
  • ERP29
  • Antigen name:
  • endoplasmic reticulum Protein 29
  • endoplasmic reticulum Protein 29
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • KLH conjugated synthetic peptide derived from human ERp29.
  • KLH conjugated synthetic peptide derived from human ERp29.
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 26 kDa
  • 26 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BIRBORB5193-100
    ORB5193-100
    100 µg
    100 µG
       
    Log in for availability
    1 * 100 µG
    1 * 100 µG

     : ORB5193-100
     : BIRBORB5193-100
     
     
    Specifications
    Antigen symbol
    ERP29
    Antigen name
    endoplasmic reticulum Protein 29
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Immunogen
    KLH conjugated synthetic peptide derived from human ERp29.
    Form
    liquid
    Molecular weight
    26 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB5193-200
    ORB5193-200
    200 µg
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG

     : ORB5193-200
     : BIRBORB5193-200
     
     
    Specifications
    Antigen symbol
    ERP29
    Antigen name
    endoplasmic reticulum Protein 29
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Immunogen
    KLH conjugated synthetic peptide derived from human ERp29.
    Form
    liquid
    Molecular weight
    26 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice