Anti-IRAK4 Mouse Monoclonal Antibody [clone: 2D3]
ABNOH00051135-M04
: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-1 Receptor-associated Kinase 4
- Antigen symbol:IRAK4
- Clonality:Monoclonal
- Clone:2D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51135
- Antigen synonyms:REN64|IRAK-4|Interleukin-1 receptor-associated kinase 4|NY-REN-64|Renal carcinoma antigen NY-REN-64|IPD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
Type: Primary
Antigen: IRAK4
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human