Anti-LMO2 Mouse Monoclonal Antibody [clone: OTI1G10]
ORIGTA506132
: OriGene
- Antibody type:Primary
- Antigen name:LIM domain only 2 (rhombotin-like 1)
- Antigen symbol:LMO2
- Clonality:Monoclonal
- Clone:OTI1G10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:18.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150
Type: Primary
Antigen: LMO2 (LIM domain only 2 (rhombotin-like 1))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human