Anti-PSMA2 Mouse Monoclonal Antibody [clone: OTI3E2]
Catalog # ORIGTA505475
Supplier: OriGene
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) subunit, Alpha Type 2
- Antigen symbol:PSMA2
- Clonality:Monoclonal
- Clone:OTI3E2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:50 to 234
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:25.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: PSMA2 (proteasome (prosome, macropain) subunit, alpha type, 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human