Anti-B3GNT2 Mouse Monoclonal Antibody [clone: OTI3A9]
ORIGTA505283
: OriGene
- Antibody type:Primary
- Antigen name:UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
- Antigen symbol:B3GNT2
- Clonality:Monoclonal
- Clone:OTI3A9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:45.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: B3GNT2 (UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human