Anti-SNAI2 Mouse Monoclonal Antibody [clone: OTI1H10]
ORIGTA504929
: OriGene
- Antibody type:Primary
- Antigen name:snail family zinc finger 2
- Antigen symbol:SNAI2
- Clonality:Monoclonal
- Clone:OTI1H10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:29.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:200-1:1000
Type: Primary
Antigen: SNAI2 (snail family zinc finger 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human