Anti-IGFBP2 Mouse Monoclonal Antibody [clone: OTI1A7]
ORIGTA504413
: OriGene
- Antibody type:Primary
- Antigen name:Insulin-Like Growth Factor Binding Protein 2
- Antigen symbol:IGFBP2
- Clonality:Monoclonal
- Clone:OTI1A7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100
Type: Primary
Antigen: IGFBP2 (insulin-like growth factor binding protein 2, 36kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human