Northern Ireland
Order Entry
Anti-NPTN Mouse Monoclonal Antibody [clone: OTI2D4]
Anti-NPTN Mouse Monoclonal Antibody [clone: OTI2D4]
Catalog # ORIGTA504330
Supplier:  OriGene
undefined
Anti-NPTN Mouse Monoclonal Antibody [clone: OTI2D4]
Catalog # ORIGTA504330
Supplier:  OriGene
Price
£582.00
Log in for availability
Unit of Measure 1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neuroplastin
  • Antigen symbol:
    NPTN
  • Clonality:
    Monoclonal
  • Clone:
    OTI2D4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    28.7 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NPTN (neuroplastin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: