Anti-FBXO21 Mouse Monoclonal Antibody [clone: OTI6C10]
ORIGTA503996
: OriGene
- Antibody type:Primary
- Antigen name:F-box protein 21
- Antigen symbol:FBXO21
- Clonality:Monoclonal
- Clone:OTI6C10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:72.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: FBXO21 (F-box protein 21)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human